Learn More
Techne™ Stuart™ 96-well Low Profile PCR Plates
Stuart™ 96-well Low Profile PCR Plates have shorter wells than the standard profile plates, decreasing the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
Brand:  Techne™ 10125-27
 View more versions of this product
Product Code. 13078523
  • 325.00 € / Pack of 25
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low profile design decreases dead space between the lid of the thermal cycler, increasing PCR efficiency
  • Available in Black, White and Clear
  • Non-sterile
Specifications
Specifications
Standard
Clear
Non-sterile
Low
8 x 12
96
25
25 Pk.