Learn More
Techne™ Stuart™ 96-well Low Profile PCR Plates
Stuart™ 96-well Low Profile PCR Plates have shorter wells than the standard profile plates, decreasing the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
Brand: Techne™ 16210-34
This item is not returnable.
View return policy
Description
- Low profile design decreases dead space between the lid of the thermal cycler, increasing PCR efficiency
- Available in Black, White and Clear
- Non-sterile
Specifications
Standard | |
White | |
Non-sterile | |
Low |
8 x 12 | |
96 | |
25 | |
25 Pk. |