Learn More
Techne™ Non-skirted 96-well PCR Plates
These non-skirted PCR plates fit the blocks of most thermal cyclers and real-type PCR instruments, they are not suitable for robotic use.
Brand:  Techne™ 99967-89
Product Code. 13008533
  • 325.00 € / Pack of 25
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low profile
  • Fits most blocks of thermal cyclers
  • Not suitable for robotic instruments
Specifications
Specifications
Standard
Clear
96
Low
8 x 12
96-well PCR Plate
25
25 Pk.