Learn More
Cole-Parmer™ Stuart™ Accessories Set
Improve and expand the performance of you vortex mixer
Brand:  Cole-Parmer™ 04729-07
Product Code. 11311990
  • 107.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with the variable-speed vortexer.
Kit contains:
  • 1x plastic cradle for standard microtitre plate
  • 1x circular foam insert for beakers and flasks (up to 500 mL capacity)
  • 1x rectangular foam insert (holds 8 x 0.2 mL, 8 x 0.5 mL and 16 x 1.5 mL microcentrifuge tubes)
  • 1x solid rectangular foam insert (blank for custom holes)
Please note: The microtitre plate holder SA8/1 should not be used at speeds above 800 rpm.
Specifications
Specifications
SA8 Vortex Mixer