Learn More
Cole-Parmer™ Drum for DR-200 Series Rotating Shaker System Drives
Optimize performance of your DR-200 Series Rotating Shaker System with interchangeable drums.
Brand:  Cole-Parmer™ 51600-99
 View more versions of this product
Product Code. 11456548
  • 1090.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
Drum
For changing your rotator's capacity
1.5 mL Microcentrifuge Tubes
240
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products