Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Optimize performance of your DR-200 Series Rotating Shaker System with this Clip Plate for Rotator Drums.
Brand:  Cole-Parmer™ 51600-93
 View more versions of this product
Product Code. 12053324
  • 190.00 € / Pack of 2
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
Tube Rack
8
19 mm
Tubes (19 mm)
For 8 Tubes