missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ ST-200-MP Stuart Three Position Magnetic Stirrer

ST-200-MP Stuart Three Position Magnetic Stirrer has three independent stirring positions yet only measures 23-5/8 in. L x 10-5/8 in. W (60 x 27 cm).

Brand:  Cole-Parmer™ 51400-02

1593.62 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.


Product Code. 11426538

  • 1765.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Powerful magnets for strong magnetic coupling and stirring capacity
  • Easily accommodate a 2-L beaker in each position
  • Stainless steel top plate does not produce eddy current to ensure powerful stirring action

Three independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.

Specifications

Specifications

100 to 1500 rpm
230 V
550 x 210 mm
4.3 in.
23.6 in.
10.6 in.
230 V, 50 Hz, 150 W
EN 61326-1:2006, EN 61010-1:2001, EN 61010-2-051:2003
SB161-3
15.4 lb.
3
150 W
60 x 27 x 11 cm
11 cm
60 cm
27 cm
50 Hz
3 x 25 mm PTFE Coated Stir Bars
Magnetic Stirrer
7 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.