missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ SBS40 Shaking Waterbath

Customize with platforms and racks

Brand:  Cole-Parmer™ 12152-02

Product Code. 12007254

  • 3190.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Choice of platforms produces a reciprocating shaking, orbital shaking, or nonshaking system
  • Analog controls and digital temperature display simplify operation
  • Overtemperature and low-level cutoffs improve safety
  • Shaking speed: 20 to 130 rpm
  • Stroke length/orbit size: 3/4 in. (20 mm)
Specifications

Specifications

Shaking Water Bath
LED
95°C
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.