missing translation for 'onlineSavingsMsg'
Learn More

Techne™ Stuart™ TE-10D Circulator

Suitable for most routine laboratory applications

Brand:  Techne™ 01262-07

Product Code. 12767929

  • 2000.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Techne™ first introduced their classic clip-on thermoregulator back in 1948, and now offer this and three other similar units operating over different temperature ranges, all designed to be used with Techne™ unheated water baths or any other suitable laboratory vessel. They will all heat, circulate and safely control the temperature of the liquid in the bath within precisely controlled limits
  • Manufacturer part number FTE10DDC
  • Temperature range -40°C to 120°C (a separate cooling coil is required for sub-ambient temperatures)
  • Excellent temperature stability: ±0.01°C at 40°C
  • Four-digit setting with a bright LED digital temperature display
  • Suitable for most routine laboratory applications
  • User adjustable over-temperature cut-out
  • Low liquid level cut-out as standard
Specifications

Specifications

-40°C to +120°C
Circulator
120/240 V
120/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.