Learn More
Cole-Parmer™ RE3022C Diaphragm Vacuum Pump
The RE3022C Diaphragm Vacuum Pump offers chemical compatibility ideal for rotary evaporator application.
Brand: Cole-Parmer™ 99966-96
3762.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
- Solid PTFE and FFKM construction resists most solvents
- Regulate vacuum level using adjustment knob
- Integral catch pots prevent liquid ingress
- 3 year warranty
- Previous part number RE3022C
Use this compact diaphragm vacuum pump for chemical laboratory vacuum generation, especially well suited to rotary evaporation. Vacuum regulator with gauge and adjustment knob allows you to create ideal vacuum conditions in the lab. Oilless pump with integrated moisture protection is low maintenance.
Specifications
PTFE | |
Vacuum Pump | |
230 V |
For Any Applications Requiring Vacuum Generation and Is Ideal For Use With The Stuart Rotary Evaporator | |
For Any Applications Requiring Vacuum Generation and Is Ideal For Use With The Stuart Rotary Evaporator |