missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 0.9060g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass Bottle |
Linear Formula | C6H5CH=CH2 |
Refractive Index | 1.5460 to 1.5480 (20°C, 589nm) |
Quantity | 1 L |
Beilstein | 05, 474 |
Merck Index | 15, 8990 |
Stabilizer | 10 to 20ppm p-tert-butylcatechol |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467569 | Certificate of Analysis | 21/02/2025 | 132790010,132790100,132790025,10103190,132790250 |
A0467570 | Certificate of Analysis | 12/02/2025 | 132790010,132790100,132790025,10103190,132790250 |
A0464803 | Certificate of Analysis | 26/07/2024 | 132790010,132790100,132790025,132790250 |
A0464821 | Certificate of Analysis | 25/07/2024 | 132790010,132790100,132790025,132790250 |
A0455059 | Certificate of Analysis | 25/01/2024 | 132790010,132790100,132790025,132790250 |
Safety and Handling



Product Identifier
- Styrene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction