missing translation for 'onlineSavingsMsg'
Learn More

Styrene-d8, for NMR, 98+% atom D, stabilized, Thermo Scientific Chemicals

111.56 € - 365.82 €

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10125113
View Documents Promotion Details
Thermo Scientific Acros
175390010
1 g Glass Bottle
111.56 €
1g
Please to purchase this item. Need a web account? Register with us today!
10072360
View Documents Promotion Details
Thermo Scientific Acros
175390050
5 g Glass Bottle
365.82 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
Specifications

Chemical Identifiers

19361-62-7
112.201
PPBRXRYQALVLMV-GDALLCCDSA-N
88025
C=CC1=CC=CC=C1
C8H8
MFCD00044231
styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene

Specifications

Liquid
0.9700g/mL
32°C
0.05% max. (Coulometric)
50.0°C to 51.0°C (25.0mmHg)
C6D5CD=CD2
styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
C=CC1=CC=CC=C1
112.201
112.22
Styrene-d8, 98+ atom% D
Colorless
98.0% min.
1.5443
19361-62-7
C8H8
MFCD00044231
PPBRXRYQALVLMV-GDALLCCDSA-N
1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
88025
NMR
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 242-995-6

missing translation for 'tsca' : TSCA

Product Identifier
  • Styrene-d8, stabilized

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains 4-tert-Butyl catechol. May produce an allergic reaction.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only