missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SUB-GEL Teaching Horizontal Gel Tank Package

SUB-GEL Teaching gel tanks are ideal for demonstration of horizontal gel electrophoresis, helps teachers to demonstrate horizontal gel electrophoresis to many students using just single gel tank and power supply.

Brand:  Fisherbrand™ FTGT6XMINI

 View more versions of this product

Product Code. 15321357

  • 394.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • High quality injection moulded construction
  • Leak free ‘Plug-and-Go’ casting dams
  • UV transparent gel trays
  • Height adjustable combs
  • Low gel and buffer volumes
  • Cassette-style electrodes - difficult to break and easy to change
  • Each tank is available with or without power supply
Specifications

Specifications

Twelve x 8 sample, 1mm MC (2 combs per gel)
Demonstrating Horizontal Gel Electrophoresis to Students
7 x 7 cm (2.7 x 2.7 in.)
96 samples with standard comb option, 196 samples using optional 16 sample combs
39.5 x 23 x 9 cm (15.5 x 9 x 3.5 in.)
Maximum Gel throughput: Six 70mm x 70mm mini Gels
Teaching Gel Tank, 6 UV trays
Six 70mm x 70mm UV trays, twelve 8 sample combs, twelve casting dams, loading guides and cables
1200 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.