missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Substance P

Sensory neuropeptide; inflammatory mediator

Brand:  Tocris Bioscience™ 1156/5

182.38 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18074152

  • 233.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Substance P is a sensory neuropeptide and inflammatory mediator. Exerts excitatory effects on central and peripheral neurons, constricts bronchioles and is involved in pain transmission.
Specifications

Specifications

95%
Store at -20°C
5 mg
33507-63-0
M.W. (Observed): 1347.6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers