Learn More
R&D Systems™ Suc-Leu-Leu-Val-Tyr-AMC (Suc-LLVY-AMC)
Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome
Brand:  R&D Systems™ S-280-05M
128.72 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18340128
  • 169.00 € / 5mg
Explore more special offers
Description
Description
  • Purity >95% by TLC, HPLC
  • Structure confirmed by NMR
Specifications
Specifications
Suc-LLVY-AMC, C40H53N5O10
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.