Learn More
Succimer, MedChemExpress
MedChemExpress Succimer is a widely used chelating agent for the treatment of Pb poisoning.
Brand: Medchem Express HY-B1768
This item is not returnable.
View return policy
Description
Succimer is a widely used chelating agent for the treatment of Pb poisoning.Specifications
Succimer | |
White | |
C4H6O4S2 | |
Dimercaptosuccinic acid DMSA | |
O=C(O)[C@@H](S)[C@@H](S)C(O)=O | |
182.22 | |
Research |
304-55-2 | |
Research | |
100 mg | |
DMSO : ≥ 100 mg/mL (548.79 mM) | |
182.22 | |
95.0% | |
Solid |
Research purposes only