missing translation for 'onlineSavingsMsg'
Learn More

Succinate, 0.2M buffer soln., pH 6.5, Thermo Scientific™

50.05 € - 65.63 €

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002789
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15414529
View Documents
Thermo Scientific Alfa Aesar
J62078.AK
250 mL
50.05 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15424529
View Documents
Thermo Scientific Alfa Aesar
J62078.AP
500 mL
65.63 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

150-90-3
162.052
ZDQYSKICYIVCPN-UHFFFAOYSA-L
CHEBI:63675
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
C4H4Na2O4
MFCD00002789
9020
disodium;butanedioate

Specifications

150-90-3
0.2 M
Liquid
Odorless
ZDQYSKICYIVCPN-UHFFFAOYSA-L
disodium;butanedioate
9020
Succinate buffer soln.
Colorless
6.5
C4H4Na2O4
MFCD00002789
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
162.052
CHEBI:63675
Videos
Safety and Handling

Safety and Handling

H335

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.