missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid 99%, Thermo Scientific Chemicals

31.26 € - 620.09 €

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10003753
SDS View Documents
Thermo Scientific Acros
158742500
250 g Plastic bottle
31.26 €
250g
Please to purchase this item. Need a web account? Register with us today!
10407000
SDS View Documents
Thermo Scientific Acros
158745000
500 g Plastic bottle
51.36 €
500g
Please to purchase this item. Need a web account? Register with us today!
10144243
SDS View Documents
Thermo Scientific Acros
158740010
1 kg Plastic bottle
106.53 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10573131
SDS View Documents
Thermo Scientific Acros
158740025
2.5 kg Plastic bottle
168.84 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11985031
SDS View Documents
Thermo Scientific Acros
158740100
10 kg Plastic drum
620.09 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinic Acid 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.