missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals
27.94 € - 62.01 €
Chemical Identifiers
CAS | 150-90-3 |
---|---|
Molecular Formula | C4H4Na2O4 |
Molecular Weight (g/mol) | 162.052 |
MDL Number | MFCD00002790 |
InChI Key | ZDQYSKICYIVCPN-UHFFFAOYSA-L |
Synonym | disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 |
PubChem CID | 9020 |
ChEBI | CHEBI:63675 |
IUPAC Name | disodium;butanedioate |
SMILES | C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10491821
|
Thermo Scientific Acros
208741000 |
100 g | Plastic bottle |
27.94 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636442
|
Thermo Scientific Acros
208745000 |
500 g | Plastic bottle |
62.01 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-90-3 | |
162.052 | |
ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
9020 | |
disodium;butanedioate |
C4H4Na2O4 | |
MFCD00002790 | |
disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
CHEBI:63675 | |
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Specifications
150-90-3 | |
White | |
Authentic | |
C4H4Na2O4 | |
MFCD00002790 | |
15, 8808 | |
Solubility in water: 300g/L (25°C). | |
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
162.052 | |
CHEBI:63675 | |
0.99 | |
Succinic acid, disodium salt, 0.99 |
>300.0°C | |
0.3% max. (120°C) | |
0.99 | |
NaO2CCH2CH2CO2Na | |
02, 606 | |
disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
disodium;butanedioate | |
9020 | |
162.05 | |
Crystalline Powder |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 205-778-7
missing translation for 'rtecsNumber' : WM7751000
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Succinic acid, disodium salt, anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only