missing translation for 'onlineSavingsMsg'
Learn More

Succinonitrile, 99+%

Product Code. 10742981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10742981

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-61-2 | C4H4N2 | 80.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-61-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00001949
InChI Key IAHFWCOBPZCAEA-UHFFFAOYSA-N
Synonym succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
PubChem CID 8062
IUPAC Name butanedinitrile
SMILES N#CCCC#N

Specifications

Melting Point 57°C
Color Colorless
Boiling Point 265°C to 267°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2CH2CN
Quantity 500 g
Beilstein 02, 615
Fieser 04,241
Merck Index 15, 9003
Solubility Information Solubility in water: 12.8%. Other solubilities: soluble in acetone,chloroform and dioxane,slightly soluble in ethanol,benzene,ether and,carbon disulfide
Formula Weight 80.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Succinonitrile, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.