Learn More
Paragon Scientific Sucrose Brix/RI Standards, Paragon Scientific™
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
Brand: Paragon Scientific SS60
This item is not returnable.
View return policy
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Primary level refractive index standards traceable to National Standards
- Manufactured in accordance with ICUMSA Methods
- Supplied in tamper evident packs with 6 weeks shelf life
Specifications
60.00 Brix | |
Colorless | |
Sucrose |
15 mL | |
Odorless | |
Liquid |
Nominal values quoted