missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, Crystal, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15598234
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.59kg
Formula Weight 342.3
Grade Multi-Compendial/NF
Physical Form Crystals
Chemical Name or Material Sucrose
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'