missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyridazine sodium, 97%, Thermo Scientific™

Product Code. 15512333 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15512333

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 23282-55-5
Molecular Formula C10H8ClN4NaO2S
Molecular Weight (g/mol) 306.7
InChI Key GHQJXOBLWKFNAD-UHFFFAOYSA-N
PubChem CID 87077948
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide;sodium
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=NN=C(C=C2)Cl.[Na]

Specifications

Melting Point 262.0°C to 270.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 306.7
Percent Purity 97%
Chemical Name or Material Sulfachloropyridazine sodium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.