missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazine, 99%, Thermo Scientific Chemicals

Product Code. 10695082 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10695082

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-68-1
Molecular Formula C12H14N4O2S
Molecular Weight (g/mol) 278.33
MDL Number MFCD00006066
InChI Key ASWVTGNCAZCNNR-UHFFFAOYSA-N
Synonym sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin
PubChem CID 5327
ChEBI CHEBI:102265
IUPAC Name 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
SMILES CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C

Specifications

Melting Point 197.0°C to 200.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9047
Formula Weight 278.32
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethazine, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.