missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxypyridazine, Thermo Scientific Chemicals

Product Code. 11916871 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11916871

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-35-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.3
InChI Key VLYWMPOKSSWJAL-UHFFFAOYSA-N
Synonym sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
PubChem CID 5330
ChEBI CHEBI:102516
IUPAC Name 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
SMILES COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 182°C to 183°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Merck Index 15,9051
Solubility Information Solubility in water: insoluble
Formula Weight 280.3
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethoxypyridazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfamethoxypyridazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.