missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic acid, ≥99.5% (alkalimetric), Honeywell™
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 2.5 kg
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
Residue after Ignition | ≤0.02% |
Quantity | 6 x 2.5 kg |
Linear Formula | NH2SO3H |
UN Number | UN2967 |
Formula Weight | 97.09 |
Percent Purity | ≥99.5% (alkalimetric) |
Chemical Name or Material | Sulfamic Acid |
Safety and Handling
Product Identifier
- Sulphamic acid
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Sulphamic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction