Learn More
Sulfanilic acid, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Linear Formula | (H2N)C6H4SO3H |
Quantity | 100 g |
Formula Weight | 173.19 |
Percent Purity | ≥98.5% (Acid) |
Physical Form | Liquid |
Chemical Name or Material | Sulfanilic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0442207 | Certificate of Analysis | 30/01/2025 | 150711000,150710010,150710025 |
A0466811 | Certificate of Analysis | 15/10/2024 | 150710010,150711000,150710025 |
A0463062 | Certificate of Analysis | 22/05/2024 | 150710010,150711000,150710025 |
A0449946 | Certificate of Analysis | 09/02/2023 | 150710100,150710010,150711000,150710025 |
A0370875 | Certificate of Analysis | 15/12/2022 | 150710100,150710010,150711000,150710025 |
Safety and Handling

- Sulfanilic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.