missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole, 98+%, Thermo Scientific Chemicals

Product Code. 11479592
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase and it inhibits folic acid synthesis in prokaryotes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfathiazole is a short-acting antimicrobial topical sulfa drug. It blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase and it Inhibits folic acid synthesis in prokaryotes. It will act against Gram positive, Gram negative and Chlamydial bacteria. It is also used as a common oral and topical antimicrobial agent.

Solubility
Slightly soluble in water, deuterated dimethyl sulfoxide and ethanol. Soluble in acetone, dilute mineral acids, potassium hydroxide solution, sodium hydroxide solution, ammonia water, dimethyl sulphoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-14-0
Molecular Formula C9H9N3O2S2
Molecular Weight (g/mol) 255.31
MDL Number MFCD00005319
InChI Key JNMRHUJNCSQMMB-UHFFFAOYSA-N
Synonym sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan
PubChem CID 5340
ChEBI CHEBI:9337
IUPAC Name 4-amino-N-(1,3-thiazol-2-yl)benzenesulfonamide
SMILES NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1

Specifications

Melting Point 200°C to 203°C
Assay Percent Range 99%
Quantity 500 g
Merck Index 14,8943
Solubility Information Slightly soluble in water,deuterated dimethyl sulfoxide and ethanol. Soluble in acetone,dilute mineral acids,potassium hydroxide solution,sodium hydroxide solution,ammonia water,dimethyl sulphoxide and methanol.
Formula Weight 255.32
Percent Purity 99%
Chemical Name or Material Sulfathiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'