missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format

Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.

Brand:  Thermo Scientific™ 24510

383.80 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10391314

  • 404.00 € / 500mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Sulfo-NHS is pure N-hydroxysulfosuccinimide, the essential compound needed to prepare water-soluble, amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.

Highlights:

  • Forms an acyl amino ester with carboxyl groups when used with the EDC
  • Sulfo-NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
  • Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
  • Sulfo-NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
  • Reactive groups: Sulfo-NHS
  • Reactive toward: amino groups (upon activation of carboxylates)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Carboxyl
Sulfo-NHS
No
No
Pierce™
Heterobifunctional
Short (<10 Å)
500 mg
Standard, Single-use, Premium-grade
Crosslinker
No
217.13
Upon receipt store at 4°C.
Ambient
Chemical Labeling
Carbodiimide
Powder
Water
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.