Learn More
Thermo Scientific™ Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.
Brand: Thermo Scientific™ A39269
173.85 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Thermo Scientific™ Pierce Sulfo-NHS is pure N-hydroxysulfosuccinimide, the essential compound needed to prepare water-soluble, amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.
- Chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods
- Efficiency of EDC-mediated coupling is increased in the presence of Sulfo-NHS
- Amine-reactive NHS esters or Sulfo-NHS esters can be made with any carboxyl-containing molecule
- Sulfo-NHS derivatives are usually directly water-soluble (can be added directly to physiologic buffers) and membrane-impermable (can be used for cell surface labeling)
- High-purity, crystalline Sulfo-NHS can be used to create high-quality activated derivatives

Specifications
Amine-Carboxyl | |
Sulfo-NHS | |
No | |
Upon receipt store at 4°C. | |
Ambient | |
Chemical Labeling | |
Sulfo-NHS Ester | |
Solid | |
Water | |
Yes |
No | |
217.13 | |
Short | |
No | |
No-Weigh™ | |
Heterobifunctional | |
Short (<10 Å) | |
10 x 2 mg | |
Standard, Single-use, Premium-grade | |
Crosslinker |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZK402935 | Certificate of Analysis | 17/12/2024 | A39269 |
ZJ401887 | Certificate of Analysis | 16/12/2024 | A39269 |
ZJ402735 | Certificate of Analysis | 16/12/2024 | A39269 |
ZJ399929 | Certificate of Analysis | 06/11/2024 | A39269 |
ZH399018 | Certificate of Analysis | 28/08/2024 | A39269 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.