missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur, colloidal powder, Honeywell™
Shop All Honeywell Fluka Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
missing translation for 'unitSize'
1kg
Specifications
Specifications
| CAS | 7704-34-9 |
| Physical Form | Colloidal Powder |
| Quantity | 1 kg |
| Molecular Formula | S |
| MDL Number | MFCD00085316 |
| UN Number | 1350 |
| Flash Point | 207°C (404.6°F) |
| Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
| InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
| SMILES | [S] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction