missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, flowers, 99%, Thermo Scientific Chemicals

22.71 € - 141.71 €

Specifications

Boiling Point 444°C
Melting Point 117°C to 120°C
CAS 7704-34-9
Physical Form Solid
Molecular Formula S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389236
View Documents
Thermo Scientific Alfa Aesar
A17260.22
100 g
22.71 €
100g
Please to purchase this item. Need a web account? Register with us today!
15493147
View Documents
Thermo Scientific Alfa Aesar
A17260.36
500 g
37.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
11399236
View Documents
Thermo Scientific Alfa Aesar
A17260.0E
2500 g
141.71 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

444°C
7704-34-9
S
117°C to 120°C
Solid
99%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulphur, flowers

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sulphur

SDS
Documents

Documents

Product Certifications