missing translation for 'onlineSavingsMsg'
Learn More

Sulfur pieces, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

92.66 € - 1219.07 €

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11385778
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010343.14
25 g
92.66 €
25g
Please to purchase this item. Need a web account? Register with us today!
11375778
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010343.22
100 g
283.41 €
100g
Please to purchase this item. Need a web account? Register with us today!
11365778
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010343.36
500 g
1219.07 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
sulfur
S
MFCD00085316
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
CHEBI:26833
[S]

Specifications

7704-34-9
Pieces
S
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
[S]
32.06
CHEBI:26833
Weak
Sulphur
Yellow
99.9995% (Metals basis)
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
248°C (478°F)
Pale yellow pieces
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III

missing translation for 'einecsNumber' : 231-722-6

missing translation for 'rtecsNumber' : WS4250000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only