Learn More
Sulfur powder, precipitated, 99.5%, Thermo Scientific Chemicals
54.87 € - 110.55 €
Chemical Identifiers
CAS | 7704-34-9 |
---|---|
Molecular Formula | S |
Molecular Weight (g/mol) | 32.06 |
MDL Number | MFCD00085316 |
InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
PubChem CID | 5362487 |
ChEBI | CHEBI:26833 |
IUPAC Name | sulfur |
SMILES | [S] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396877
|
Thermo Scientific Alfa Aesar
010785.36 |
500 g |
54.87 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386877
|
Thermo Scientific Alfa Aesar
010785.A3 |
2 kg |
110.55 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7704-34-9 | |
32.06 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
5362487 | |
sulfur |
S | |
MFCD00085316 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
CHEBI:26833 | |
[S] |
Specifications
7704-34-9 | |
99.50% | |
MFCD00085316 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
248°C (478°F) | |
2.07g/cm3 |
Yellow | |
S | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
[S] | |
32.06 | |
CHEBI:26833 | |
Weak | |
Sulphur |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-722-6
missing translation for 'rtecsNumber' : WS4250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sulphur powder, precipitated
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sulphur
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.