missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, precipitated, Honeywell Fluka™

Product Code. 15611370
Click to view available options
Quantity:
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., USP, Puriss., precipitated, 99.5-100.5%,

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Quantity 2.5 kg
UN Number 1350
Flash Point 207°C (404.6°F)
Solubility Information In accordance with carbon disulfide, 1g/5mL
Formula Weight 32.07g/mol
Percent Purity 99.5 to 100.5%
Grade puriss.
Ignition Point 450°F
Residue after Ignition ≤0.2% (as SO4)
Vapor Pressure 1mmHg (183.8°C), 10mmHg (246°C)
Density Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'