missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Technical, Flowers, SLR, Fisher Chemical™

Product Code. 10469760
Click to view available options
Quantity:
1 kg
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445°C
Melting Point 113°C
Color Yellow
Physical Form Solid
Quantity 3 kg
Formula Weight 32.06g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Sulfur flowers
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.