Learn More
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
44.60 € - 310.69 €
Specifications
Chemical Name or Material | Sulfuric acid |
---|---|
Color | Colorless |
Name Note | 0.1N (0.05M) Standard Solution |
CAS | 7664-93-9, 7732-18-5 |
Concentration | 0.0998 to 0.1002N (20°C) |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10183550
|
Thermo Scientific Acros
124230010 |
1 L | Plastic Bottle |
44.60 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10473055
|
Thermo Scientific Acros
124230025 |
2.5 L | Plastic Bottle |
82.60 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13458328
|
Thermo Scientific Acros
124230250 |
25 L | Plastic jerrycan | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sulfuric acid | |
0.1N (0.05M) Standard Solution | |
0.0998 to 0.1002N (20°C) | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07 |
Colorless | |
7664-93-9, 7732-18-5 | |
H2O4S | |
Liquid | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Pure |
Safety and Handling
GHS H Statement:
May be corrosive to metals.
GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Sulfuric acid, 0.1N (0.05M) standard solution
RUO – Research Use Only