missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.2N Standardized Solution, Thermo Scientific Chemicals

32.96 € - 144.72 €

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492117
View Documents
Thermo Scientific Alfa Aesar
045062.K2
1 L
32.96 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15402127
View Documents
Thermo Scientific Alfa Aesar
045062.K7
4 L
62.01 €
4 L
Please to purchase this item. Need a web account? Register with us today!
15412127
View Documents
Thermo Scientific Alfa Aesar
045062.M4
20 L
144.72 €
20 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sulfuric acid, 0.2N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
OS(O)(=O)=O
H2O4S
MFCD00064589
sulfuric acid

Specifications

7664-93-9
Liquid
Miscible with water.
OS(O)(=O)=O
98.07
Sulfuric acid, 0.2N Standardized Solution
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
98.08
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-639-5

missing translation for 'rtecsNumber' : WS5600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.