missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid, 75% v/v aq. soln. Product Code.: 15481817

Sulfuric acid, 75% v/v aq. soln.

Product Code. 15481817
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15481817

Brand: Thermo Scientific Alfa Aesar 042555.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid has an useful applications in domestic acidic drain cleaner, as a drying agent, electrolyte in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacturing, oil refining, waste water processing, and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide, shows an useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Colorless to pale yellow liquid
CAS 7664-93-9, 7732-18-5
Odor Acrid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 75% v/v aqueous
Quantity 1 L
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Miscible with water,acetic acid and alcohol.
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 84.7%; Water: 15.3%
Chemical Name or Material Sulfuric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, 75% v/v aq. soln. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.