missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals Product Code.: 10056612

Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals

Product Code. 10056612
1 L, Plastic Bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10056612

Brand: Thermo Scientific Acros 133610011

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Melting Point 10.0°C
Boiling Point 290.0°C
Density 1.8300g/mL
Packaging Plastic Bottle
Specific Gravity 1.83
Assay Percent Range 96%
Quantity 1 L
Viscosity 21 mPa.s (25°C)
Linear Formula H2SO4
Physical Form Oily Liquid
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Percent Purity 96%
Grade Extra Pure
Chemical Name or Material Sulfuric acid, 96% Solution in Water
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB00Y5683 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number133610026,133610250,133610010,133610011,133610025
Lot NumberA0463034 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number133610026,133610250,133610010,133610011,133610025
Lot NumberA0443568 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number133610026,133610250,133610010,133610011,133610025
Lot NumberA0458701 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number133610026,133610250,133610010,133610011,133610025
Lot NumberB00V0389 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number133610026,133610250,133610010,133610011,133610025
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.