missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals Product Code.: 11341608

Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals

Product Code. 11341608
10 mL
Click to view available options
Quantity:
10 mL
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11341608

Brand: Thermo Scientific Alfa Aesar 011000.AB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, insecticides, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless to Yellow
Concentration ≤92%
Density 1.84 g/mL
Odor Acrid
Quantity 10 mL
Physical Form Liquid
UN Number UN1830
Merck Index 14,8974
Solubility Information Sulfuric acid is miscible with water at all concentrations.
Formula Weight 98.08
Percent Purity 99.9999%
Assay (metals basis)
Chemical Name or Material Sulfuric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.