missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS, 95.0-98.0%

Product Code. 11337588
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
4 x 1 kg
missing translation for 'unitSize'
1kg
4kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11337588

missing translation for 'mfr': Thermo Scientific Alfa Aesar 033273.D6

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.84
Assay Percent Range 95.0 to 98.0%
Quantity 4 x 1 kg
Physical Form Liquid
Solubility Information Sulfuric acid is miscible with water at all concentrations.
Formula Weight 98.08
Grade ACS
Chemical Name or Material Sulfuric Acid
compliance-icons
Product Identifier
  • Sulfuric acid, ACS, 95.0-98.0%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.