missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals

29.15 € - 226.00 €

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 7
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10624792
SDS View Documents
Thermo Scientific Acros
124645000
500 mL Glass Bottle
30.95 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10699824
SDS View Documents
Thermo Scientific Acros
124645001
500 mL Plastic bottle
29.15 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10244790
SDS View Documents
Thermo Scientific Acros
124640010
1 L Glass Bottle
45.23 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10765663
SDS View Documents
Thermo Scientific Acros
124640011
1 L Plastic bottle
45.93 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10011240
SDS View Documents
Thermo Scientific Acros
124640025
2.5 L Glass Bottle
88.14 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10322963
SDS View Documents
Thermo Scientific Acros
124640026
2.5 L Plastic bottle
91.25 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11904091
SDS View Documents
Thermo Scientific Acros
124640100
10 L Plastic drum
226.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-639-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.