missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid concentrate, For 1 L standard solution, 0.5 M H2SO4 (1.0 N), Honeywell Fluka™

Product Code. 15675280
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Packaging:
Ampoule
Plastic Bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

for 1 L standard solution, 0.5 M H2SO4 (1.0 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Especificaciones

Packaging Ampoule
Quantity 1 Ea.
Linear Formula H2SO4
UN Number UN2796
Beilstein 2037554
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid concentrate
compliance-icons
Nombre del producto
  • Sulfuric acid concentrate
Palabra de advertencia
  • Danger
Clasificación
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'