missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals

1203.00 €

Chemical Identifiers

CAS 8014-95-7
Molecular Formula H2O7S2
Molecular Weight (g/mol) 178.13
MDL Number MFCD00137792
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES O=S(=O)=O.OS(O)(=O)=O
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10679453
View Documents
Thermo Scientific Acros
419970025
2.5 kg Glass bottle
1203.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8014-95-7
178.13
HIFJUMGIHIZEPX-UHFFFAOYSA-N
H2O7S2
MFCD00137792
O=S(=O)=O.OS(O)(=O)=O

Specifications

Brown to Colorless
2.0°C
1.9200g/mL
1.92
H2SO4·SO3
MFCD00137792
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
HIFJUMGIHIZEPX-UHFFFAOYSA-N
sulfuric acid;sulfur trioxide
24681
20 to 30% (free SO3)
Sulfuric acid
8014-95-7
138.0°C
0.01% max.
H2O7S2
Liquid
15, 9104
Solubility in water: miscible. Other solubilities: soluble in ethanol
O=S(=O)=O.OS(O)(=O)=O
178.13
178.14
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'rtecsNumber' : WS5605000

Product Identifier
  • Sulfuric acid, fuming, approx. 20% free SO3

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.