missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
HDPE Plastic Bottle
Description
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Specifications
Specifications
pH | 1 |
Color | Colorless |
Melting Point | 10°C |
Boiling Point | 290°C |
Packaging | HDPE Plastic Bottle |
Vapor Pressure | 1mmHg 146 |
Quantity | 1 L |
Viscosity | 21mPa at 25°C |
Physical Form | Liquid |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'