missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Min 95% d=1.83, Technical, Fisher Chemical

Product Code. 10496880
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

pH 1
Color Colorless
CAS 7664-93-9
Melting Point 10°C
Boiling Point 290°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg 146
Quantity 2.5 L
Molecular Formula H2O4S
Viscosity 21mPa at 25°C
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.07g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'