missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss., meets analytical spec. of BP, Ph. Eur., 95-97%, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
50 kg
6 x 1 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Unit Size:
1 L
2.5 L
5 L
50kg
Pack of 4
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 310°C |
| CAS Min % | ≥50.0000%, ≥1.0000% |
| CAS Max % | ≤100.0000%, <5.0000% |
| Density | 1.840 g/cm3 |
| Packaging | Plastic Bottle |
| Quantity | 6 x 1 L |
| Linear Formula | H2SO4 |
| UN Number | UN1830 |
| Beilstein | 2037554 |
| Formula Weight | 98.08 |
| Show More |
Safety and Handling
Product Identifier
- Sulfuric acid
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction