missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
50 kg
6 x 1 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 310°C |
CAS Min % | ≥50.0000%, ≥1.0000% |
CAS Max % | ≤100.0000%, <5.0000% |
Density | 1.840 g/mL at 25 °C(lit.) |
Packaging | Metal Drum |
Residue after Ignition | ≤5ppm |
Vapor Pressure | 1mmHg (146°C) |
Quantity | 50 kg |
Linear Formula | H2SO4 |
UN Number | UN1830 |
Show More |
Safety and Handling
Product Identifier
- Sulfuric acid
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction