missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Description
H2O4S, CAS Number-7664-93-9, acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil, 6AMP, 100 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072
Specifications
Specifications
pH | 1.5 |
Color | Colorless |
Boiling Point | 100°C |
Packaging | Ampoule |
Vapor Pressure | 1mmHg at 20°C |
Quantity | 1 Ampule |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Sulfuric acid 0.05M (0.1N) solution (0.5M (1N) Concentrate)
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : Sulfuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction